Gene Transcripts

Gene Symbol
PRAMEF5
Entrez Gene ID
343068
Gene Name
PRAME family member 5
Gene Aliases
PRAMEF23, PRAMEF5L
Chromosome Location
Chr. 1: 13254198 - 13263435 on Build GRCh38
UniGene ID
Hs.567864
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013407.2 NP_001013425.2 4 1670

View Full Product Details