Gene Transcripts

Gene Symbol
GP9
Entrez Gene ID
2815
Gene Name
glycoprotein IX platelet
Gene Aliases
CD42a, GPIX
Chromosome Location
Chr. 3: 129055449 - 129062411 on Build GRCh38
UniGene ID
Hs.1144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000174.4 NP_000165.1 3 466
XM_005247374.3 XP_005247431.1 5 762
XM_011512701.1 XP_011511003.1 6 844
XM_011512702.1 XP_011511004.1 5 719
GenBank mRNA BC030229.1 - 3 431
M25827.1 - 1 211
X52997.1 - 3 466

View Full Product Details