Gene Transcripts

Gene Symbol
GPR3
Entrez Gene ID
2827
Gene Name
G protein-coupled receptor 3
Gene Aliases
ACCA
Chromosome Location
Chr. 1: 27392644 - 27395814 on Build GRCh38
UniGene ID
Hs.66542
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005281.3 NP_005272.1 2 973
GenBank mRNA AK291185.1 - 2 960
AK314868.1 - 2 952
BC032702.1 - 2 969 5247608
HQ709189.1 - 1 874

View Full Product Details