Gene Transcripts
- Gene Symbol
- DPPA3P2
- Entrez Gene ID
- 400206
- Gene Name
- developmental pluripotency associated 3 pseudogene 2
- Gene Aliases
- STELLAR
- Chromosome Location
- Chr. 14: 36371318 - 36372369 on Build GRCh38
- UniGene ID
- Hs.528118
- Species
- Homo sapiens
- Gene Symbol
- DPPA3
- Entrez Gene ID
- 359787
- Gene Name
- developmental pluripotency associated 3
- Gene Aliases
- STELLA
- Chromosome Location
- Chr. 12: 7711454 - 7717559 on Build GRCh38
- UniGene ID
- Hs.131358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_199286.3 |
NP_954980.1
|
3, 4 |
497 |
|
GenBank mRNA |
AY317075.1 |
- |
3, 4 |
497 |
|
View Full Product Details