Gene Transcripts

Gene Symbol
DDTL
Entrez Gene ID
100037417
Gene Name
D-dopachrome tautomerase-like
Gene Aliases
KB-226F1.2
Chromosome Location
Chr. 22: 23966221 - 23972559 on Build GRCh38
UniGene ID
Hs.744172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001084393.1 NP_001077862.1 3 374
GenBank mRNA AK296110.1 - 3 324

View Full Product Details