Gene Transcripts

Gene Symbol
Chrne
Entrez Gene ID
11448
Gene Name
cholinergic receptor, nicotinic, epsilon polypeptide
Gene Aliases
AChrepsilon, Acre, nAChRE
Chromosome Location
Chr. 11: 70614883 - 70619835 on Build GRCm38
UniGene ID
Mm.4980
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009603.1 NP_033733.1 4 294
GenBank mRNA AF032863.1 - 3 147
X55718.1 - 4 294

View Full Product Details