Gene Transcripts
- Gene Symbol
- Chrne
- Entrez Gene ID
- 11448
- Gene Name
- cholinergic receptor, nicotinic, epsilon polypeptide
- Gene Aliases
- AChrepsilon, Acre, nAChRE
- Chromosome Location
- Chr. 11: 70614883 - 70619835 on Build GRCm38
- UniGene ID
- Mm.4980
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009603.1 |
NP_033733.1
|
2 |
114 |
|
GenBank mRNA |
X55718.1 |
- |
2 |
114 |
|
View Full Product Details