Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009634.6 | NP_033764.2 | 4 | 488 | |
GenBank mRNA | AK049372.1 | - | 4 | 460 | |
AK051111.1 | - | 4 | 460 | ||
AK152092.1 | - | 4 | 460 | ||
AK166019.1 | - | 4 | 456 | ||
AK168906.1 | - | 4 | 453 | ||
AK171725.1 | - | 4 | 469 | ||
AK201011.1 | - | 3 | 150 | ||
BC020187.1 | - | 4 | 464 | 4913637 | |
U20225.1 | - | 4 | 464 |