Gene Transcripts
- Gene Symbol
- Aif1
- Entrez Gene ID
- 11629
- Gene Name
- allograft inflammatory factor 1
- Gene Aliases
- AI607846, AIF-1, D17H6S50E, G1, Iba1
- Chromosome Location
- Chr. 17: 35170991 - 35176087 on Build GRCm38
- UniGene ID
- Mm.10747
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019467.2 |
NP_062340.1
|
6, 7 |
503 |
|
GenBank mRNA |
AB013745.1 |
- |
4, 5 |
230 |
|
AF074959.1 |
- |
4, 5 |
241 |
|
AK006184.1 |
- |
6, 7 |
503 |
|
AK006562.1 |
- |
6, 7 |
504 |
|
BC021539.1 |
- |
4, 5 |
371 |
5346103 |
CT010265.1 |
- |
4, 5 |
189 |
|
CT010385.1 |
- |
4, 5 |
189 |
|
D86382.1 |
- |
4, 5 |
266 |
|
EF101555.1 |
- |
3, 4 |
141 |
|
View Full Product Details