Gene Transcripts

Gene Symbol
Ank1
Entrez Gene ID
11733
Gene Name
ankyrin 1, erythroid
Gene Aliases
Ank-1, nb, pale
Chromosome Location
Chr. 8: 22974836 - 23150502 on Build GRCm38
UniGene ID
Mm.334444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110783.3 NP_001104253.1 28 3485
NM_001277280.2 NP_001264209.1 27 3461
NM_001277281.2 NP_001264210.1 27 3182
NM_001277284.2 NP_001264213.1 27 3182
NM_001277286.2 NP_001264215.1 28 3471
NM_001277289.2 NP_001264218.1 28 3471
NM_031158.4 NP_112435.2 28 3471
GenBank mRNA AK147278.1 - 28 3440
AK147483.1 - 28 3467
AK147597.1 - 28 3419
AK158131.1 - 27 3454
BC079910.1 - 27 3153 30664024
BC138029.1 - 28 3205 8861048
BC145102.1 - 28 3205
BC171944.1 - 28 3205 9054230
M84756.1 - 27 3159
X69063.1 - 29 3455
X69065.1 - 16 1853

View Full Product Details