Gene Transcripts

Gene Symbol
Ap1m2
Entrez Gene ID
11768
Gene Name
adaptor protein complex AP-1, mu 2 subunit
Gene Aliases
D9Ertd818e, [m]1B, mu1B
Chromosome Location
Chr. 9: 21294613 - 21312349 on Build GRCm38
UniGene ID
Mm.22239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110300.1 NP_001103770.1 8 1060
NM_009678.2 NP_033808.2 8 1054
GenBank mRNA AF067146.1 - 8 1028
AK010478.1 - 8 1039
AK082674.1 - 917
BC003704.1 - 8 993 3488231

View Full Product Details