Gene Transcripts

Gene Symbol
Aqp6
Entrez Gene ID
11831
Gene Name
aquaporin 6
Gene Aliases
AI790337
Chromosome Location
Chr. 15: 99599053 - 99605477 on Build GRCm38
UniGene ID
Mm.202309
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175087.4 NP_780296.1 3 615
GenBank mRNA AK082699.1 - 3 616
BC115586.1 - 3 616 40102369
BC115587.1 - 3 615
DQ464060.1 - 3 616
DQ826418.1 - 3 607

View Full Product Details