Gene Transcripts

Gene Symbol
Atp2a1
Entrez Gene ID
11937
Gene Name
ATPase, Ca++ transporting, cardiac muscle, fast twitch 1
Gene Aliases
SERCA1
Chromosome Location
Chr. 7: 126445860 - 126463103 on Build GRCm38
UniGene ID
Mm.35134
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007504.2 NP_031530.2 13 1610
GenBank mRNA AY081946.1 - 13 1641
BC036292.1 - 13 1610 4191741

View Full Product Details