Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000855.2 | NP_000846.1 | 7 | 2295 | |
NM_001256424.1 | NP_001243353.1 | 8 | 2388 | ||
GenBank mRNA | BC130484.1 | - | 7 | 2001 | |
BC130488.1 | - | 7 | 2001 | 40146519 | |
BC144033.1 | - | 8 | 2064 | 9052553 | |
X63282.1 | - | 7 | 2295 | ||
Z50053.1 | - | 8 | 2388 |