Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284508.1 | NP_001271437.1 | 4 | 653 | |
NM_031161.4 | NP_112438.1 | 3 | 490 | ||
GenBank mRNA | AK002677.1 | - | 3 | 458 | |
BC028487.1 | - | 4 | 598 | 1400830 | |
M11739.1 | - | 2 | 229 |