Gene Transcripts
- Gene Symbol
- Ccne1
- Entrez Gene ID
- 12447
- Gene Name
- cyclin E1
- Gene Aliases
- AW538188, CycE1
- Chromosome Location
- Chr. 7: 38097984 - 38107490 on Build GRCm38
- UniGene ID
- Mm.16110
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007633.2 |
NP_031659.2
|
4, 5 |
377 |
|
GenBank mRNA |
AK089950.1 |
- |
4, 5 |
309 |
|
AK163046.1 |
- |
4, 5 |
366 |
|
AK165658.1 |
- |
4, 5 |
367 |
|
BC062152.1 |
- |
4, 5 |
404 |
6510163 |
BC084588.1 |
- |
4, 5 |
421 |
6506897 |
BC106191.1 |
- |
4, 5 |
377 |
6336006 |
BC138660.1 |
- |
3, 4 |
210 |
8861680 |
BC138662.1 |
- |
3, 4 |
210 |
8861683 |
X75888.1 |
- |
3, 4 |
217 |
|
View Full Product Details