Gene Transcripts

Gene Symbol
GYPB
Entrez Gene ID
2994
Gene Name
glycophorin B (MNS blood group)
Gene Aliases
CD235b, GPB, GYP, MNS, PAS-3, SS
Chromosome Location
Chr. 4: 143995172 - 144021860 on Build GRCh38
UniGene ID
Hs.654368
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304382.1 NP_001291311.1 5 327
NM_002100.5 NP_002091.3 4 247
XM_011531903.2 XP_011530205.1 4 382
XM_011531904.2 XP_011530206.1 5 2886
XM_011531905.2 XP_011530207.1 5 534
XM_017008137.1 XP_016863626.1 4 2581
GenBank mRNA AY950609.1 - 4 189
AY950610.1 - 4 189
AY950611.1 - 4 189
AY950612.1 - 5 282
BC069310.1 - 4 239 7262104
BC121077.1 - 4 223 40119713
BC121078.1 - 4 223 40119716
EU338223.1 - 3 127
EU338224.1 - 5 350
EU338225.1 - 5 319
EU338226.1 - 5 284
EU338228.1 - 3 127
EU338229.1 - 5 350
EU338235.1 - 3 127
EU338236.1 - 5 350
EU338237.1 - 6 443
EU338238.1 - 2 88
J02982.1 - 4 199
X08055.1 - 6 325

View Full Product Details