Gene Transcripts

Gene Symbol
Cdk4
Entrez Gene ID
12567
Gene Name
cyclin-dependent kinase 4
Gene Aliases
Crk3
Chromosome Location
Chr. 10: 127063537 - 127067288 on Build GRCm38
UniGene ID
Mm.6839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009870.3 NP_034000.1 6, 7 853
GenBank mRNA AK003804.1 - 6, 7 820
AK011871.1 - 6, 7 841
AK012064.1 - 6, 7 822
AK013400.1 - 6, 7 791
AK019380.1 - 6, 7 806
AK078440.1 - 6, 7 807
AK146107.1 - 6, 7 820
AK153465.1 - 6, 7 853
AK218927.1 - 2, 3 140
BC031599.1 - 5, 6 1529
BC046336.1 - 6, 7 853 6312184
BC052694.1 - 6, 7 848 5714971
L01640.1 - 5, 6 680

View Full Product Details