Gene Transcripts

Gene Symbol
Cdx1
Entrez Gene ID
12590
Gene Name
caudal type homeobox 1
Gene Aliases
Cdx, Cdx-1
Chromosome Location
Chr. 18: 61018862 - 61036199 on Build GRCm38
UniGene ID
Mm.144448
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009880.3 NP_034010.3 3 713
GenBank mRNA BC019986.1 - 3 713
M37163.1 - 3 563

View Full Product Details