Gene Transcripts

Gene Symbol
Coro1a
Entrez Gene ID
12721
Gene Name
coronin, actin binding protein 1A
Gene Aliases
Clabp, Lmb3, TACO, p57
Chromosome Location
Chr. 7: 126699774 - 126704816 on Build GRCm38
UniGene ID
Mm.290482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301374.1 NP_001288303.1 8 1130
NM_009898.3 NP_034028.1 8 1043
GenBank mRNA AF047388.1 - 7 877
AF143955.1 - 8 1014
AF495468.1 - 8 981
AK040885.1 - 8 987
AK075799.1 - 8 978
AK146290.1 - 8 981
AK150962.1 - 8 985
AK151755.1 - 8 985
AK155527.1 - 8 986
AK155860.1 - 8 981
AK165296.1 - 8 985
AK172447.1 - 8 981
BC002136.1 - 8 971 3484185
BC053398.1 - 8 961 30076939

View Full Product Details