Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007737.2 | NP_031763.2 | 51 | 4170 | |
GenBank mRNA | AK084285.1 | - | 13 | 987 | |
AK084538.1 | - | 10 | 954 | ||
AK084803.1 | - | 51 | 3808 | ||
AK132413.1 | - | 51 | 3952 | ||
AK139130.1 | - | 51 | 3745 | ||
AK147220.1 | - | 51 | 3952 | ||
AK147328.1 | - | 51 | 4171 | ||
AK151929.1 | - | 51 | 3954 | ||
AK160008.1 | - | 51 | 3744 | ||
BC043696.1 | - | 25 | 1985 | ||
BC055077.1 | - | 51 | 3921 | 30059991 | |
L02918.1 | - | 51 | 3802 |