Gene Transcripts

Gene Symbol
Ncan
Entrez Gene ID
13004
Gene Name
neurocan
Gene Aliases
C230035B04, Cspg3, Cspg3-rs, Tgfbit
Chromosome Location
Chr. 8: 70093082 - 70120870 on Build GRCm38
UniGene ID
Mm.268079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007789.3 NP_031815.2 11 3376
GenBank mRNA AK082298.1 - 1 101
BC065118.1 - 11 3342 6853253
X84727.1 - 11 3338

View Full Product Details