Gene Transcripts

Gene Symbol
Cyp2c68
Entrez Gene ID
433247
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 68
Gene Aliases
9030012A22Rik
Chromosome Location
Chr. 19: 39688836 - 39741101 on Build GRCm38
UniGene ID
Mm.335660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039555.2 NP_001034644.2 6 963
GenBank mRNA AK136413.1 - 5 593

Gene Symbol
Cyp2c40
Entrez Gene ID
13099
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 40
Gene Aliases
AI265721, AI662255
Chromosome Location
Chr. 19: 39767073 - 39812814 on Build GRCm38
UniGene ID
Mm.353875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010004.2 NP_034134.2 6 995
GenBank mRNA AF047727.1 - 6 909
BC139471.1 - 6 995 8862493
BC139472.1 - 6 995 8862494

Gene Symbol
Cyp2c69
Entrez Gene ID
100043108
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 69
Gene Aliases
AI098658
Chromosome Location
Chr. 19: 39842660 - 39886769 on Build GRCm38
UniGene ID
Mm.482331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104525.1 NP_001097995.1 6 965
GenBank mRNA BC145630.1 - 7 986

View Full Product Details