Gene Transcripts
- Gene Symbol
- Daxx
- Entrez Gene ID
- 13163
- Gene Name
- Fas death domain-associated protein
- Gene Aliases
- Chromosome Location
- Chr. 17: 33909381 - 33915590 on Build GRCm38
- UniGene ID
- Mm.271809
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199733.1 |
NP_001186662.1
|
6, 7 |
2215 |
|
NM_007829.4 |
NP_031855.3
|
6, 7 |
2115 |
|
GenBank mRNA |
AF006040.1 |
- |
5, 6 |
1966 |
|
AK090356.1 |
- |
7, 8 |
1926 |
|
AK133863.1 |
- |
6, 7 |
2023 |
|
AK144332.1 |
- |
6, 7 |
2048 |
|
AK145906.1 |
- |
6, 7 |
2046 |
|
AK146743.1 |
- |
6, 7 |
2042 |
|
AK151279.1 |
- |
6, 7 |
2036 |
|
AK153149.1 |
- |
6, 7 |
2003 |
|
AK156425.1 |
- |
6, 7 |
2213 |
|
AK172616.1 |
- |
6, 7 |
2216 |
|
AY734459.1 |
- |
4, 5 |
1679 |
|
AY734464.1 |
- |
1, 2 |
288 |
|
AY734465.1 |
- |
4, 5 |
1590 |
|
AY734466.1 |
- |
|
1870 |
|
AY758274.1 |
- |
1, 2 |
421 |
|
BC128373.1 |
- |
6, 7 |
2025 |
40111681 |
KJ650207.1 |
- |
5, 6 |
1942 |
|
View Full Product Details