Gene Transcripts

Gene Symbol
Eci1
Entrez Gene ID
13177
Gene Name
enoyl-Coenzyme A delta isomerase 1
Gene Aliases
Dci, eci
Chromosome Location
Chr. 17: 24426683 - 24439316 on Build GRCm38
UniGene ID
Mm.291743
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010023.4 NP_034153.2 7 750
GenBank mRNA AK004838.1 - 7 751
AK029481.1 - 7 751
AK077386.1 - 7 741
BC022712.1 - 7 734 4487371
BC054444.1 - 7 737 6476161
Z14049.1 - 7 730

View Full Product Details