Gene Transcripts
- Gene Symbol
- Adam28
- Entrez Gene ID
- 13522
- Gene Name
- a disintegrin and metallopeptidase domain 28
- Gene Aliases
- C130072N01Rik, D430033C21Rik, Dtgn1, MDC-L, MDC-Lm, MDC-Ls, MDCL, TECADAM, eMDCII
- Chromosome Location
- Chr. 14: 68604994 - 68655887 on Build GRCm38
- UniGene ID
- Mm.117450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048175.2 |
NP_001041640.1
|
4, 5 |
472 |
|
NM_010082.2 |
NP_034212.1
|
4, 5 |
472 |
|
NM_183366.3 |
NP_899222.1
|
4, 5 |
472 |
|
GenBank mRNA |
AF153350.1 |
- |
4, 5 |
379 |
|
AF163290.1 |
- |
4, 5 |
472 |
|
AF163291.1 |
- |
4, 5 |
472 |
|
AF163292.1 |
- |
4, 5 |
472 |
|
AF163293.1 |
- |
4, 5 |
472 |
|
AK036800.1 |
- |
4, 5 |
412 |
|
AK052479.1 |
- |
4, 5 |
363 |
|
BC058782.1 |
- |
4, 5 |
347 |
6310763 |
View Full Product Details