Gene Transcripts

Gene Symbol
Edar
Entrez Gene ID
13608
Gene Name
ectodysplasin-A receptor
Gene Aliases
ED1R, ED3, ED5, EDA-A1R, EDA3, dl
Chromosome Location
Chr. 10: 58600780 - 58675696 on Build GRCm38
UniGene ID
Mm.174523
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010100.3 NP_034230.1 11 1241
GenBank mRNA AF160502.1 - 11 1241
AK004576.2 - 5 370
BC068315.1 - 11 1199 30098537

View Full Product Details