Gene Transcripts

Gene Symbol
Epha1
Entrez Gene ID
13835
Gene Name
Eph receptor A1
Gene Aliases
5730453L17Rik, AL033318, Eph, Esk
Chromosome Location
Chr. 6: 42358487 - 42373310 on Build GRCm38
UniGene ID
Mm.133330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023580.4 NP_076069.2 3 481
GenBank mRNA AF131197.1 - 3 423
AK017662.1 - 3 449
AK028478.1 - 3 482
BC071215.1 - 3 445 4196138

View Full Product Details