Gene Transcripts

Gene Symbol
Ephb6
Entrez Gene ID
13848
Gene Name
Eph receptor B6
Gene Aliases
Cekl, Mep
Chromosome Location
Chr. 6: 41605040 - 41620507 on Build GRCm38
UniGene ID
Mm.271976
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146351.1 NP_001139823.1 11, 12 2414
NM_007680.4 NP_031706.3 11, 12 2410
GenBank mRNA AK076442.1 - 11, 12 2237
AK087423.1 - 11, 12 2325
AK163823.1 - 2, 3 132
BC031924.1 - 10, 11 2022 4911959
L77867.1 - 11, 12 2104

View Full Product Details