Gene Transcripts
- Gene Symbol
- Ercc3
- Entrez Gene ID
- 13872
- Gene Name
- excision repair cross-complementing rodent repair deficiency, complementation group 3
- Gene Aliases
- BTF2 p89, Ercc-3, XPB
- Chromosome Location
- Chr. 18: 32240305 - 32270151 on Build GRCm38
- UniGene ID
- Mm.282335
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133658.1 |
NP_598419.1
|
12, 13 |
2020 |
|
GenBank mRNA |
AK151188.1 |
- |
12, 13 |
2025 |
|
AK152347.1 |
- |
12, 13 |
2025 |
|
AK152988.1 |
- |
12, 13 |
2032 |
|
AK154745.1 |
- |
12, 13 |
2026 |
|
AK160187.1 |
- |
6, 7 |
1124 |
|
BC016595.1 |
- |
12, 13 |
1991 |
3153847 |
BC022956.1 |
- |
11, 12 |
2124 |
|
BC024446.1 |
- |
3, 4 |
366 |
|
BC026575.1 |
- |
12, 13 |
2002 |
4955015 |
S71186.1 |
- |
12, 13 |
2020 |
|
View Full Product Details