Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007997.1 | NP_032023.1 | 9 | 914 | |
GenBank mRNA | AK134068.1 | - | 9 | 905 | |
AK183932.1 | - | 2 | 73 | ||
BC057146.1 | - | 9 | 848 | ||
BC071268.1 | - | 9 | 914 | 6517167 | |
D49920.1 | - | 9 | 914 |