Gene Transcripts
- Gene Symbol
- Lpin1
- Entrez Gene ID
- 14245
- Gene Name
- lipin 1
- Gene Aliases
- 4631420P06, Kiaa0188, Lipin1, fld, mKIAA0188
- Chromosome Location
- Chr. 12: 16535669 - 16646968 on Build GRCm38
- UniGene ID
- Mm.153625
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130412.1 |
NP_001123884.1
|
11, 12 |
1745 |
|
NM_015763.4 |
NP_056578.2
|
11, 12 |
1700 |
|
NM_172950.3 |
NP_766538.2
|
10, 11 |
1601 |
|
GenBank mRNA |
AB093214.1 |
- |
6, 7 |
920 |
|
AF180471.1 |
- |
10, 11 |
1580 |
|
AF412811.1 |
- |
10, 11 |
1638 |
|
AK014526.1 |
- |
10, 11 |
1661 |
|
AK019539.1 |
- |
11, 12 |
1838 |
|
AK030921.1 |
- |
10, 11 |
1604 |
|
AK154837.1 |
- |
11, 12 |
1742 |
|
BC042462.1 |
- |
10, 11 |
1654 |
4211202 |
View Full Product Details