Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008027.2 | NP_032053.1 | 6, 7 | 641 | |
GenBank mRNA | AF145044.1 | - | 5, 6 | 482 | |
AK150692.1 | - | 6, 7 | 663 | ||
AK150827.1 | - | 6, 7 | 590 | ||
AK167323.1 | - | 6, 7 | 568 | ||
AY167925.1 | - | 6, 7 | 647 | ||
BC004647.1 | - | 6, 7 | 651 | 3158071 | |
U90435.1 | - | 6, 7 | 641 |