Gene Transcripts

Gene Symbol
G6pd2
Entrez Gene ID
14380
Gene Name
glucose-6-phosphate dehydrogenase 2
Gene Aliases
G6pdx-ps1, Gpd-2, Gpd2
Chromosome Location
Chr. 5: 61808843 - 61810477 on Build GRCm38
UniGene ID
Mm.347430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019468.2 NP_062341.2 1 220
GenBank mRNA BC120827.1 - 1 220
BC137684.1 - 1 220 8860703

View Full Product Details