Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162477.1 | NP_001155949.1 | 2 | 239 | |
NM_010248.2 | NP_034378.2 | 2 | 239 | ||
GenBank mRNA | AB018414.1 | - | 2 | 164 | |
AF104244.1 | - | 2 | 239 | ||
BC103524.1 | - | 2 | 214 | 40039291 | |
BC103525.1 | - | 2 | 214 | 40039280 | |
BC103526.1 | - | 2 | 214 | 40039281 | |
BC103527.1 | - | 2 | 214 | 40039282 |