Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038010.2 | NP_001033099.1 | 18 | 2401 | |
NM_020004.5 | NP_064388.2 | 18 | 2404 | ||
GenBank mRNA | AF254441.1 | - | 18 | 2341 | |
AK145205.1 | - | 16 | 2401 | ||
AK147042.1 | - | 18 | 2349 | ||
AK150225.1 | - | 18 | 2382 | ||
AK154866.1 | - | 17 | 2479 | ||
AK155347.1 | - | 18 | 2578 | ||
AK155791.1 | - | 18 | 2403 | ||
AK155842.1 | - | 18 | 2402 | ||
AK158079.1 | - | 18 | 2392 | ||
BC003983.1 | - | 3 | 199 | ||
BC011088.1 | - | 18 | 2311 | ||
BC051952.1 | - | 16 | 2625 | ||
BC060050.1 | - | 16 | 2625 | ||
BC063752.1 | - | 18 | 2386 | 6333090 |