Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016905.2 | NP_058601.2 | 6 | 970 | |
GenBank mRNA | AB027012.1 | - | 6 | 926 | |
AK010340.1 | - | 6 | 967 | ||
AK011103.1 | - | 6 | 971 | ||
AK045694.1 | - | 6 | 944 | ||
AK088151.1 | - | 6 | 971 | ||
AK168716.1 | - | 6 | 930 | ||
BC016602.1 | - | 6 | 970 | 3490224 | |
BC050151.1 | - | 6 | 941 | 30024030 |