Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021332.2 | NP_067307.2 | 9 | 914 | |
GenBank mRNA | AJ001692.1 | - | 9 | 904 | |
BC103624.1 | - | 9 | 903 | ||
BC105649.1 | - | 9 | 903 | ||
BC105650.1 | - | 9 | 903 | ||
BC139464.1 | - | 9 | 941 | 8862486 | |
BC139466.1 | - | 9 | 941 | 8862488 | |
BC145626.1 | - | 9 | 941 | 9054194 | |
DQ093397.1 | - | 9 | 914 | ||
DQ093398.1 | - | 9 | 914 |