Gene Transcripts

Gene Symbol
Gnaq
Entrez Gene ID
14682
Gene Name
guanine nucleotide binding protein, alpha q polypeptide
Gene Aliases
1110005L02Rik, 6230401I02Rik, AA408290, AW060788, Dsk1, Dsk10, Galphaq, Gq, GqI
Chromosome Location
Chr. 19: 16132831 - 16387453 on Build GRCm38
UniGene ID
Mm.439701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008139.5 NP_032165.3 4 1060
GenBank mRNA AK147393.1 - 4 1061
BC057583.1 - 4 800 6408959
M55412.1 - 4 601

View Full Product Details