Gene Transcripts

Gene Symbol
Gp1bb
Entrez Gene ID
14724
Gene Name
glycoprotein Ib, beta polypeptide
Gene Aliases
Chromosome Location
Chr. 16: 18620319 - 18622403 on Build GRCm38
UniGene ID
Mm.475622
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001999.1 NP_001001999.1 1 2000
NM_010327.2 NP_034457.1 2 1762
GenBank mRNA AA222119.1 - 1 449
BC138156.1 - 1 1954 8861175
BC145160.1 - 1 1954 9053711

View Full Product Details