Gene Transcripts

Gene Symbol
Rhpn1
Entrez Gene ID
14787
Gene Name
rhophilin, Rho GTPase binding protein 1
Gene Aliases
BB023497, Grbp, Rhophilin, mKIAA1929
Chromosome Location
Chr. 15: 75704285 - 75714419 on Build GRCm38
UniGene ID
Mm.57052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163465.1 NP_001156937.1 3 335
NM_008164.2 NP_032190.2 3 335
GenBank mRNA AK143132.1 - 3 316
AK220431.1 - 3 252
BC004843.1 - 3 305
BC052010.1 - 3 294 5717786
U43194.1 - 3 229

View Full Product Details