Gene Transcripts

Gene Symbol
PSMB7
Entrez Gene ID
5695
Gene Name
proteasome subunit beta 7
Gene Aliases
Z
Chromosome Location
Chr. 9: 124353465 - 124415473 on Build GRCh38
UniGene ID
Hs.213470
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002799.3 NP_002790.1 5 504
GenBank mRNA AJ420455.1 - 2 182
AK098757.1 - 5 473
AK223007.1 - 5 457
AK223356.1 - 5 469
AK314302.1 - 5 473
BC000509.2 - 5 460 2822305
BC008414.1 - 5 470
BC017116.2 - 4 395
BT007218.1 - 5 456
CR456721.1 - 5 456
D38048.1 - 5 470
GU727627.1 - 5 473

View Full Product Details