Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159551.1 | NP_001153023.1 | 7 | 830 | |
NM_010376.4 | NP_034506.1 | 7 | 830 | ||
GenBank mRNA | AF483214.1 | - | 7 | 797 | |
AF515662.1 | - | 7 | 709 | ||
AJ345032.1 | - | 7 | 840 | ||
AK007664.1 | - | 7 | 812 | ||
AK012600.1 | - | 7 | 823 | ||
AK028109.1 | - | 7 | 799 | ||
AK049101.1 | - | 7 | 804 | ||
AK050664.1 | - | 7 | 773 | ||
AK080966.1 | - | 7 | 792 | ||
AK159176.1 | - | 7 | 804 | ||
AK169922.1 | - | 7 | 787 | ||
AK171790.1 | - | 6 | 632 | ||
AK186920.1 | - | 1 | 29 | ||
AK216222.1 | - | 1 | 29 | ||
BC034217.1 | - | 2 | 104 | ||
BC045195.1 | - | 3 | 186 | ||
DQ168449.1 | - | 7 | 846 |