Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172117.1 | NP_001165588.1 | 13 | 1655 | |
NM_010407.4 | NP_034537.2 | 13 | 1655 | ||
GenBank mRNA | AK149736.1 | - | 13 | 1621 | |
AK150290.1 | - | 13 | 1629 | ||
AK150709.1 | - | 13 | 1638 | ||
AK152506.1 | - | 13 | 1628 | ||
AK155975.1 | - | 13 | 1568 | ||
AK159234.1 | - | 13 | 1588 | ||
AK165315.1 | - | 13 | 1620 | ||
BC010478.1 | - | 13 | 1651 | 3978518 | |
BC013260.1 | - | 1 | 34 | ||
J03023.1 | - | 13 | 1544 | ||
Y00487.1 | - | 13 | 1550 |