Gene Transcripts

Gene Symbol
Hes2
Entrez Gene ID
15206
Gene Name
hairy and enhancer of split 2 (Drosophila)
Gene Aliases
HES-2
Chromosome Location
Chr. 4: 152158867 - 152162469 on Build GRCm38
UniGene ID
Mm.57038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008236.4 NP_032262.2 5 1173
GenBank mRNA AK053792.1 - 5 1174
BC138112.1 - 5 1501 8861131

View Full Product Details