Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008250.2 | NP_032276.1 | 1 | 870 | |
GenBank mRNA | AK154090.1 | - | 1 | 548 | |
BB664380.1 | - | 1 | 494 | ||
BC050146.1 | - | 1 | 125 | ||
BC064068.1 | - | 1 | 870 | 1380899 | |
X58250.1 | - | 1 | 986 |