Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008255.2 | NP_032281.2 | 15 | 2076 | |
GenBank mRNA | AK079302.1 | - | 3 | 275 | |
AK159899.1 | - | 14 | 1964 | ||
BC019782.1 | - | 14 | 1952 | ||
BC059873.1 | - | 6 | 757 | ||
BC085083.1 | - | 15 | 1988 | 5320486 |