Gene Transcripts

Gene Symbol
Hmox1
Entrez Gene ID
15368
Gene Name
heme oxygenase 1
Gene Aliases
D8Wsu38e, HO-1, HO1, Hemox, Hmox, Hsp32
Chromosome Location
Chr. 8: 75093618 - 75100593 on Build GRCm38
UniGene ID
Mm.276389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010442.2 NP_034572.1 2 226
GenBank mRNA AK150934.1 - 2 223
AK151005.1 - 2 223
AK151012.1 - 2 214
AK151627.1 - 2 201
AK153420.1 - 2 223
AK153516.1 - 2 223
AK153566.1 - 2 223
AK154598.1 - 2 224
AK159521.1 - 2 223
AK159870.1 - 2 227
AK159959.1 - 2 227
AK169227.1 - 2 223
BC010757.1 - 2 195 4235374
BY319630.1 - 2 228
M33203.1 - 2 168
X13356.1 - 2 168

View Full Product Details