Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010442.2 | NP_034572.1 | 3 | 580 | |
GenBank mRNA | AK150934.1 | - | 3 | 577 | |
AK151005.1 | - | 3 | 577 | ||
AK151012.1 | - | 3 | 568 | ||
AK151627.1 | - | 3 | 555 | ||
AK153420.1 | - | 3 | 577 | ||
AK153516.1 | - | 3 | 577 | ||
AK153566.1 | - | 3 | 577 | ||
AK154598.1 | - | 3 | 578 | ||
AK159521.1 | - | 3 | 577 | ||
AK169227.1 | - | 3 | 577 | ||
BC010757.1 | - | 3 | 549 | 4235374 | |
M33203.1 | - | 3 | 522 | ||
X13356.1 | - | 3 | 522 |