Gene Transcripts

Gene Symbol
Ier2
Entrez Gene ID
15936
Gene Name
immediate early response 2
Gene Aliases
AI317238, Ch1, Pip92
Chromosome Location
Chr. 8: 84661331 - 84662852 on Build GRCm38
UniGene ID
Mm.399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010499.4 NP_034629.3 1 1493
GenBank mRNA AK032692.1 - 1491
AK144601.1 - 1 1490
AK150860.1 - 1 1488
AK152747.1 - 1 1488
AK170433.1 - 1490
AK172332.1 - 1 1433
BC002067.1 - 1 1478 3590296
M31042.1 - 1 1490
M59821.1 - 1 1493

View Full Product Details